missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C1GALT1C1 (aa 164-291) Control Fragment Recombinant Protein

Codice prodotto. 30193780
Click to view available options
Quantity:
100 μL
Dimensione della confezione:
100µL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30193780

Marca: Invitrogen™ RP108717

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a type II transmembrane protein that is similar to the core 1 beta1,3-galactosyltransferase 1, which catalyzes the synthesis of the core-1 structure, also known as Thomsen-Friedenreich antigen, on O-linked glycans. This gene product lacks the galactosyltransferase activity itself, but instead acts as a molecular chaperone required for the folding, stability and full activity of the core 1 beta1,3-galactosyltransferase 1. Mutations in this gene have been associated with Tn syndrome. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifica

Accession Number Q96EU7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29071
Name Human C1GALT1C1 (aa 164-291) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500002I11Rik; AB030184; beta 1,3-galactosyltransferase 2; beta1,3-galactosyltransferase 2; C1GALT1 specific chaperone 1; C1galt1c1; C1GALT1-specific chaperone 1; C1GALT2; C1Gal-T2; C38H2-L1; C38H2-like protein 1; C81205; core 1 beta1,3-galactosyltransferase 2; core 1 beta3-galactosyltransferase-specific molecular chaperone; core 1 beta3-Gal-T2; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 2; COSMC; HSPC067; mC1Gal-T2; MST143; MSTP143; RGD1311230; TNPS; UNQ273/PRO310
Common Name C1GALT1C1
Gene Symbol C1GALT1C1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SQPFYLGHTIKSGDLEYVGMEGGIVLSVESMKRLNSLLNIPEKCPEQGGMIWKISEDKQLAVCLKYAGVFAENAEDADGKDVFNTKSVGLSIKEAMTYHPNQVVEGCCSDMAVTFNGLTPNQMHVMMY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato