missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C19orf48 Control Fragment Recombinant Protein

Product Code. 30182499
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182499

Brand: Invitrogen™ RP99147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59814 (PA5-59814. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The C19orf48 gene, located on chromosome 19q13, encodes a protein involved in various biological processes, including its role as a minor histocompatibility antigen recognized by CD8+ T cells. This gene has been implicated in oncogenesis due to its elevated expression in certain cancer types, such as breast cancer, where it has been associated with poor prognosis. The non-synonymous single-nucleotide polymorphisms within C19orf48 contribute to differential T-cell recognition in donor and recipient cells, highlighting its significance in immune responses. Additionally, C19orf48 has been linked with SNORD88C, a small nucleolar RNA (snoRNA) that guides methylation of rRNA, further illustrating its involvement in cellular proliferation and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84798
Name Human C19orf48 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C19orf48; chromosome 19 open reading frame 48; multidrug resistance-related protein; Uncharacterized protein C19orf48
Common Name C19orf48
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MTVLEAVLEIQAITGSRLLSMVPGPARPPGSCWDPTQCTRTWLLSHTPRRRWISGLPR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.