missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C17orf96 (aa 2-38) Control Fragment Recombinant Protein

Product Code. 30209799
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209799

Brand: Invitrogen™ RP106610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65930 (PA5-65930. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Elongin BC and Polycomb repressive complex 2-associated protein (EPOP) also know as C17ORF96 is a scaffold protein that serves as a bridging partner between the PRC2/EED-EZH2 complex and the elongin BC complex. It is required to fine tune the transcriptional status of polycomb group (PcG) target genes in embryonic stem cells (ESCs). C17ORF96 also plays a key role in genomic regions that display both active and repressive chromatin properties in pluripotent stem cells by sustaining low level expression of PcG target genes. It acts by recruiting the elongin BC complex, thereby restricting excessive activity of the PRC2/EED-EZH2 complex. C17ORF96 also has interacts with USP7 and promotes deubiquitination of H2B at promoter sites.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A6NHQ4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 100170841
Name Human C17orf96 (aa 2-38) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA409164; AI413509; AW539173; C17orf96; C9H17orf96; chromosome 17 open reading frame 96; CUNH17orf96; E13; E130012A19Rik; elongin BC and polycomb repressive complex 2 associated protein; elongin BC and Polycomb repressive complex 2-associated protein; Embryonic stem cell-specific PRC2 subunit p48; EPOP; ES cell-specific PRC2 subunit p48; esPRC2p48; hypothetical protein LOC691153; LOC691153; proline rich 28; Proline-rich protein 28; PRR28; RIKEN cDNA E130012A19 gene; uncharacterized protein C17orf96; uncharacterized protein C17orf96 homolog; Uncharacterized protein ENSP00000317905
Common Name C17orf96
Gene Symbol EPOP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ETLCPAPRLAVPASPRGSPCSPTPRKPCRGTQEFSPL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.