missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C17orf85 (aa 147-256) Control Fragment Recombinant Protein

Product Code. 30193806
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193806

Brand: Invitrogen™ RP89980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53033 (PA5-53033. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Associates with NCBP1/CBP80 to form an alternative cap-binding complex (CBC) which plays a key role in mRNA export. NCBP3 serves as adapter protein linking the capped RNAs (m7GpppG-capped RNA) to NCBP1/CBP80. Unlike the conventional CBC with NCBP2 which binds both small nuclear RNA (snRNA) and messenger (mRNA) and is involved in their export from the nucleus, the alternative CBC with NCBP3 does not bind snRNA and associates only with mRNA thereby playing a role in only mRNA export. The alternative CBC is particularly important in cellular stress situations such as virus infections and the NCBP3 activity is critical to inhibit virus growth.
TRUSTED_SUSTAINABILITY

Spécification

Accession Number Q53F19
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55421
Name Human C17orf85 (aa 147-256) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1200014J11Rik; C11H17orf85; C130061O14Rik; C17orf85; C19H17orf85; C78393; C9H17orf85; CTD-3195I5.1; CTD-3195I5.5; ELG; ELG protein; HSA277841; Ncbp3; nuclear cap binding subunit 3; nuclear cap-binding protein subunit 3; Protein ELG; RGD1308139; RIKEN cDNA 1200014J11 gene; similar to RIKEN cDNA 1200014J11; uncharacterized protein C17orf85 homolog
Common Name C17orf85
Gene Symbol NCBP3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EYPPAHIEWLDDTSCNVVWLDEMTATRALINMSSLPAQDKIRSRDASEDKSAEKRKKDKQEDSSDDDEAEEGEVEDENSSDVELDTLSQVEEESLLRNDLRPANKLAKGN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis