missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C17orf47 Isoform 7 Control Fragment Recombinant Protein

Product Code. 30213491
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213491

Brand: Invitrogen™ RP93472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55846 (PA5-55846. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

C17orf47 Isoform 7 refers to a variant of the C17orf47 gene, located on chromosome 17 (17p13.3). This gene encodes a protein that plays a role in various cellular processes, though its exact functions are still being explored. The gene has several splice variants, with Isoform 7 being one of them, potentially impacting its functional properties. C17orf47 is expressed in multiple tissues, and its expression may vary depending on the tissue type, suggesting a role in tissue-specific cellular functions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43236
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5414
Name Human C17orf47 Isoform 7 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C17orf47; Chromosome 17 open reading frame 47; Septin 4
Common Name C17orf47 Isoform 7
Gene Symbol SEPTIN4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YSAYPETKPSAKVLVSSQVESNVRTPIRGNSEVGRRVTISPGVQSVEPTHHVTVPSVSEGSHKSSMFVTPEPIYKQQTQKPPEITYMSQGPTPRYPELSQKPSIHAEL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.