missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human C14orf80 (aa 153-241) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58637 (PA5-58637. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
C14orf80 may play a role in counteracting perturbation of actin filaments, such as after treatment with the actin depolymerizing microbial metabolite Chivosazole F.
Specifications
Specifications
| Accession Number | Q86SX3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 283643 |
| Name | Human C14orf80 (aa 153-241) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | C14orf80; chromosome 14 open reading frame 80; TEDC1; Tubulin epsilon and delta complex protein 1; uncharacterized protein C14orf80 |
| Common Name | C14orf80 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction