missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C12orf30 (aa 740-824) Control Fragment Recombinant Protein

Product Code. 30182048
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182048

Brand: Invitrogen™ RP97686

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58734 (PA5-58734. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the auxiliary subunit of the heteromeric N-terminal acetyltransferase B complex. This complex acetylates methionine residues that are followed by acidic or asparagine residues.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14CX7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80018
Name Human C12orf30 (aa 740-824) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4833422K13Rik; A630046C11; AI450868; C12orf30; C330023M02Rik; Liver regeneration-related protein LRRGT00164; LRRGT00164; MDM20; mitochondrial distribution and morphology 20; mitochondrial distribution and morphology protein 20; N(alpha)-acetyltransferase 25, NatB auxiliary subunit; Naa25; Nalpha acteyltransferase 25; N-alpha-acetyltransferase 25, NatB auxiliary subunit; NAP1; NatB complex subunit MDM20; N-terminal acetyltransferase B complex subunit MDM20; N-terminal acetyltransferase B complex subunit NAA25; p120; RGD1305685
Common Name C12orf30
Gene Symbol Naa25
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TGKRFIEKDIQYPFLGPVPTRMGGFFNSGCSQCQISSFYLVNDIYELDTSGLEDTMEIQERIENSFKSLLDQLKDVFSKCKGDLL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.