missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human c-Rel Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4606-SP
This item is not returnable.
View return policy
Description
c-Rel Monoclonal specifically detects c-Rel in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| c-Rel | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| NP_002899 | |
| REL | |
| E. coli-derived recombinant human c-Rel Met1-Val619 Accession # NP_002899 | |
| 25 μg | |
| Primary | |
| Detects human c-Rel in Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 321422 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cRel, C-Rel, C-Rel proto-oncogene protein, I-Rel, oncogene REL, avian reticuloendotheliosis, proto-oncogene c-Rel, REL, v-rel avian reticuloendotheliosis viral oncogene homolog, v-rel reticuloendotheliosis viral oncogene homolog (avian) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5966 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction