missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human C-Reactive Protein (CRP) ELISA Kit
Description
Includes
- Pre-coated ELISA plate
- Detection Antibody. Pre-titrated, HRP-conjugated antibody
- Standard. Protein for generating standard curve and calibrating samples
- Tris Wash Buffer Powder
- Substrate Solution. Tetramethylbenzidine (TMB) solution
- Stop Solution
- Certificate of Analysis. Lot-specific instructions for dilution of reagents
- Human C-reactive protein (CRP) kit contains all of the necessary buffers and reagents for performing enzyme-linked immunosorbent assay (ELISA)
- To ensure optimal performance from this ELISA kit, please only use the components included in the kit. Exchanging of components is not recommended, as a change in signal may occur
Specifications
Specifications
| Assay Sensitivity | 150pg/mL |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 96 Tests |
| Storage Requirements | 2°C to 8°C |
| Target | C-Reactive Protein (CRP) |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?