missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human C-Reactive Protein/CRP DuoSet ELISA

Product Code. 30021487
Change view
Click to view available options
Quantity:
15 x 96-well Plates
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30021487 15 x 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021487 Supplier R&D Systems™ Supplier No. DY1707

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human C-Reactive Protein/CRP DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human C-Reactive Protein/CRP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human C-Reactive Protein/CRP.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 15 x 96-well Plates
Conjugate Biotin
Assay Range 15.60 to 1000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 1401
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.