missing translation for 'onlineSavingsMsg'
Learn More

Human C-Reactive Protein/CRP Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18761249
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18761249 25 μg 25µg
18762808 100 μg 100µg
18755499 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18761249 Supplier R&D Systems Supplier No. MAB17071SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C-Reactive Protein/CRP
Applications ELISA
Classification Monoclonal
Clone 232007
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P02741
Gene Alias C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895
Gene Symbols CRP
Host Species Mouse
Immunogen Human plasma-derived CRP Accession # P02741
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1401
Test Specificity Detects human CRP in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.