missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human C-Peptide Quantikine ELISA Kit

Product Code. 18721849
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18721849

Brand: R&D Systems™ DICP00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

a 2.5 hour solid-phase ELISA designed to measure human Insulin C-Peptide in cell culture supernates, serum, plasma, and urine.

The Human C-Peptide Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human C-Peptide in cell culture supernates (100 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 28.90 - 1850 pmol/L
Assay Sensitivity 2.88 pmol/L
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and synthetic Human Insulin C-Peptide
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 3630
Sample Volume Cell Culture Supernates (100uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym C-P, IDDM2, ILPR, insulin, Insulin C-Peptide, IRDN, MODY10, proinsulin, Proinsulin C-Peptide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.