missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human c-Jun Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF2670
This item is not returnable.
View return policy
Description
c-jun Polyclonal specifically detects c-jun in Human samples. It is validated for Western Blot.
Specifications
| c-jun | |
| Polyclonal | |
| Unconjugated | |
| P05412 | |
| JUN | |
| E. coli-derived recombinant human c-Jun Thr2-Phe331 Accession # P05412 | |
| 100 μg | |
| Primary | |
| Detects human c-Jun in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Activator protein 1, AP1, AP-1, cJun, c-Jun, enhancer-binding protein AP1, JUN, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3725 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction