missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C/EBP alpha (aa 165-184) Control Fragment Recombinant Protein

Product Code. 30195253
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195253

Brand: Invitrogen™ RP107609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The C/EBP (CCAAT/enhancer binding protein) family of transcription factors--alpha, beta, delta, and epsilon)--are widely expressed proteins found in a variety of tissues. These domains contain an evolutionarily conserved five-amino acid motif (the regulatory domain motif RDM) that conforms to the consensus sequence (I/V/L) KXEP. One of the major roles of these transcription factors is that of metabolic regulation in liver and fat cells. Studies suggest that these transcription factors act in a cascade aiding in the development of the adipose cell. Three C/EBP beta isoforms (LAP*, LAP, and LIP) arise from different translation initiation and display different function in gene regulation. From a neurological standpoint, C/EBPs have been implicated in the mediation of nerve growth factor, altering synaptic plasticity, and mediation of inflammatory response in AD patients.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49715
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1050
Name Human C/EBP alpha (aa 165-184) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C EBP; C/EBP; c/EBP alpha; C/EBPa; C/ebpalpha; C/EBP-alpha; CAAT/enhancer-binding protein DNA-binding protein; CAAT/enhancer-binding protein, DNA-binding protein; CBF-A; CCAAT enhancer binding protein alpha; CCAAT/enhancer binding protein (C/EBP), alpha; CCAAT/enhancer binding protein alpha; CCAAT/enhancer binding protein, alpha; CCAAT/enhancer-binding protein alpha; Cebp; Cebpa; DBPCEP; OTTHUMP00000220269
Common Name C/EBP alpha
Gene Symbol CEBPA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence REEDEAKQLALAGLFPYQPP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.