missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTK Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5807-SP
This item is not returnable.
View return policy
Description
BTK Monoclonal specifically detects BTK in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| BTK | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.05 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q06187 | |
| BTK | |
| E. coli-derived recombinant human BTK Glu280-Val377 Accession # Q06187 | |
| 25 μg | |
| Primary | |
| Detects endogenous human BTK in Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 566928 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Agammaglobulinaemia tyrosine kinase, AGMX1, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, IMD1, PSCTK1, tyrosine-protein kinase BTK, XLA, XLAMGC126261 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 695 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction