missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Brorin/VWC2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6147-SP
This item is not returnable.
View return policy
Description
Brorin/VWC2 Monoclonal specifically detects Brorin/VWC2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Brorin/VWC2 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q2TAL6 | |
| VWC2 | |
| Mouse myeloma cell line NS0-derived recombinant human Brorin/VWC2 Ser28-Met325 Accession # Q2TAL6 | |
| 25 μg | |
| Primary | |
| Detects human Brorin/VWC2 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse Brorin/VWC2 and no cross-reactivity with recombinant human (rh) vWF-A2 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 615609 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Brain-Specific Chordin-Like, Brain-Specific Chordin-Like Protein, brorin, PSST739, UNQ739, Von Willebrand Factor C Domain Containing 2, Von Willebrand Factor C Domain-Containing Protein 2, VWC2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 375567 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction