missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Breast Whole Tissue Lysate (Adult Whole Normal)

Product Code. 18271482 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18271482 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18271482 Supplier Novus Biologicals™ Supplier No. NB82059203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Western Blot, Immunoprecipitation, In vitro assay

The Human Breast Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation, In vitro assay.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, In vitro assay
Tissue Breast
Quantity 1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Purity Protein
Test Specificity Total Protein - Human Adult Normal Tissue: Breast
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Human
Host Species Human
Concentration 4.9 mg/mL
Product Type Human Breast Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.