missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Breast Tissue Lysate (Adult Normal)

Product Code. 18451262 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18451262

Brand: Novus Biologicals™ NBP227999

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Normal Adult Tissue Lystate

TRUSTED_SUSTAINABILITY

Specifications

Preparation Method These lysates are proteomic discovery tools.Researchers should validate and optimize for individual use. Potential applications MAY include WB, immunoprecipitation, protein-protein interactions, ligand binding, ELISA. Note: For use in 1D and 2D gel electrophoresis, the addition of a denaturing gel loading buffer with reducing agents may be required.
Quantity 0.1 mg
Content And Storage Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles.
Target Breast
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.