missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BRE (aa 104-204) Control Fragment Recombinant Protein

Product Code. 30194705
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194705

Brand: Invitrogen™ RP108868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

BRCC45 was initially suggested to be a housekeeping protein that is highly expressed in brain and reproductive organs. Later experiments indicated BRCC45 forms a complex with the breast and ovarian predisposition proteins BRCA1 and BRCA2 as well as RAD51 and BRCC36. This complex has a ubiquitin E3 ligase activity and is thought to enhance cellular survival following DNA damage. BRCC45 has also been suggested to function as a death receptor-associated anti-apoptotic protein by inhibiting the BID-induced activation of the mitochondrial apoptotic pathway. Higher levels of BRCC45 were detected in the majority of hepatocellular carcinomas, suggesting that BRCC45 may promote tumorigenesis when overexpressed. At least three isoforms of BRCC45 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NXR7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9577
Name Human BRE (aa 104-204) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6030405P19Rik; AI429776; B830038C02Rik; BABAM2; brain and reproductive organ-expressed (TNFRSF1A modulator); brain and reproductive organ-expressed protein; BRCA1/BRCA2-containing complex subunit 45; BRCA1/BRCA2-containing complex, subunit 4; BRCA1-A complex subunit BRE; BRCC4; BRCC45; BRE; bre {ECO:0000312; BRISC and BRCA1-A complex member 2; RGD:735111}
Common Name BRE
Gene Symbol BABAM2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SWNPSNPECLLLVVKELVQQYHQFQCSRLRESSRLMFEYQTLLEEPQYGENMEIYAGKKNNWTGEFSARFLLKLPVDFSNIPTYLLKDVNEDPGEDVALLS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.