missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BRCAA1 (aa 729-825) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82965 (PA5-82965. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The exact function of C18orf25 remains unknown. There are two named isoforms.
Specifications
Specifications
| Accession Number | Q4LE39 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51742 |
| Name | Human BRCAA1 (aa 729-825) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 180 kDa Sin3-associated polypeptide; ARID domain-containing protein 4 B; ARID4B; AT rich interactive domain 4 B (Rbp1 like); AT rich interactive domain 4 B (RBP1-like); AT-rich interaction domain 4 B; AT-rich interactive domain-containing protein 4 B; Bcaa; BRCAA1; breast cancer-associated antigen; breast cancer-associated antigen 1; breast cancer-associated antigen BRCAA1; breast carcinoma-associated antigen; histone deacetylase complex subunit SAP180; Rb-binding protein homolog; RBBP1L1; RBP1L1; retinoblastoma binding protein 1-like 1; retinoblastoma-binding protein 1-like 1; retinoblastoma-binding protein 1-related protein; Sap180; SIN3A-associated protein 180; Sin3-associated polypeptide p180 |
| Common Name | BRCAA1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction