Learn More
Human BRCA2 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 6 publications
Brand:  R&D Systems MAB2476-SP
Product Code. 18794449
  • £179.00 / 25µg
Description
BRCA2 Monoclonal specifically detects BRCA2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BRCA2
Monoclonal
LYOPH
Western Blot 0.2 ug/mL, Immunohistochemistry 8-25 ug/mL
P51587
BRCA2
E. coli-derived recombinant human BRCA2 Met1-Thr200 Accession # P51587
25 μg
Primary
Detects endogenous human BRCA2.
Human
Purified
Western Blot, Immunohistochemistry
234403
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BRCA1/BRCA2-containing complex, subunit 2, BRCC2, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast cancer susceptibility protein BRCA2, breast cancer type 2 susceptibility protein, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1Fanconi anemia, complementation group D1, Fanconi anemia group D1 protein, GLM3, PNCA2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
675
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1