missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Brain Pons Whole Tissue Lysate (Adult Whole Multiple Sclerosis)
Shop All Bio Techne ProductsDescription
The Human Brain Pons Whole Tissue Lysate (Adult Whole Multiple Sclerosis) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Tissue | Nervous |
| Quantity | 1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Total Protein - Multiple Sclerosis Disease: Brain: Pons |
| Cellular Fraction | Whole |
| Tissue Condition | Multiple Sclerosis |
| Cross Reactivity | Human |
| Host Species | Human |
| Concentration | 5 mg/mL |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?