missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BOC/Brother of CDO ELISA Kit

Product Code. 17185279
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17185279

Brand: Invitrogen™ EH51RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human BOC/Brother of CDO quantitates human BOC/Brother of CDO in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human BOC/Brother of CDO.

CDON and BOC are cell surface receptors of the immunoglobulin repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 4732455C11; biregional Cdon binding protein; biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein; Boc; BOC cell adhesion associated, oncogene regulated; Boc homolog; Boc homolog (mouse); Brother of CDO; brother of CDON; CDON2; cell adhesion associated, oncogene regulated 2; mFLJ00376; Protein BOC; UNQ604/PRO1190
Assay Range 0.085 to 20 ng/mL
Assay Sensitivity 0.085 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 91653
Gene Symbol Boc
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target BOC
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.