missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BOC Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2036
This item is not returnable.
View return policy
Description
BOC Polyclonal specifically detects BOC in Human samples. It is validated for Western Blot.
Specifications
| BOC | |
| Polyclonal | |
| Biotin | |
| Q9BWV1 | |
| BOC | |
| Mouse myeloma cell line NS0-derived recombinant human BOC Asp31-Asp852 Accession # Q9BWV1 | |
| 50 μg | |
| Primary | |
| Detects human BOC in Western blots. In Western blots, approximately 30% cross-reactivity with recombinant mouse BOC is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Boc homolog (mouse), brother of CDO, Protein BOC | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 91653 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction