missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BMPR1A (ALK-3) Fc Chimera Recombinant Protein

Product Code. 17666535
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 17666535

missing translation for 'mfr': Invitrogen™ RP87720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

BMPR1A is a Type I member of the TGF beta receptor superfamily of transmembrane ser/thr kinases which phosphorylates intracellular SMADs in response to bone morphogenic proteins. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P36894
For Use With (Application) Control
Formulation PBS with no preservative
Gene ID (Entrez) 657
Molecular Weight (g/mol) 40.4 kDa
Name Human BMPR1A (ALK-3) Fc Chimera
Quantity 1 mg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration ≤1 EU/μg
Gene Alias 10q23del; 1110037I22Rik; activin A receptor, type II-like kinase 3; activin receptor-like kinase 3; ACVRLK3; ALK3; ALK-3; AU045487; BMP type-1 A receptor; BMP-2/BMP-4 receptor; Bmpr; Bmpr1a; BMPR-1 A; BMPRIA; BMPR-IA; Bone morphogenetic protein 4 receptor; bone morphogenetic protein receptor type 1 A; bone morphogenetic protein receptor type IA; bone morphogenetic protein receptor type-1 A; bone morphogenetic protein receptor, type 1 A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5
Common Name BMPR1A
Gene Symbol BMPR1A
Biological Activity ED50 = 120 ng/mL; determined by the dose-dependent inhibition of human BMP-4-induced alkaline phosphatase production by ATDC5 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence Recombinant human BMPR1A is a homodimer containing two 362 amino acid subunits with a C-terminal linker (IEGRMD) and a human IgG1 Fc-tag (AA Sequence: QNLDSMLHGT GMKSDSDQKK SENGVTLAPE DTLPFLKCYC SGHCPDDAIN NTCITNGHCF AIIEEDDQGE TTLASGCMKY EGSDFQCKDS PKAQLRRTIE CCRTNLCNQY LQPTLPPVVI GPFFDGSIRI EGRMDDKTHT CPPCPAPELL GGPSVFLFPP KPKDTLMISR TPEVTCVVVD VSHEDPEVKF NWYVDGVEVH NAKTKPREEQ YNSTYRVVSV LTVLHQDWLN GKEYKCKVSN KALPAPIEKT ISKAKGQPRE PQVYTLPPSR DELTKNQVSL TCLVKGFYPS DIAVEWESNG QPENNYKTTP PVLDSDGSFF LYSKLTVDKS RWQQGNVFSC SVMHEALHNH YTQKSLSLSP GK)
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade >95% by SDS-Page
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.