missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-8 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1073-SP
This item is not returnable.
View return policy
Description
BMP-8 Polyclonal specifically detects BMP-8 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BMP-8 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP8, BMP8A, BMP-8A, OP-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 353500 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| AAP74559 | |
| BMP8A | |
| E. coli-derived recombinant human BMP-8a Ala264-His402 Accession # AAP74559 | |
| 25 μg | |
| Primary | |
| Detects human BMP-8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) BMP-2, rhBMP-3, rhBMP-4, rhBMP-5, rhBMP-6 and rhBMP-7 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction