missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-7 Quantikine ELISA Kit

Product Code. 30001018
Click to view available options
Quantity:
1 Pack
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 30001018

Brand: R&D Systems™ SBP700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human BMP-7 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human BMP-7 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human BMP-7 in cell culture supernates (50 ul), bone extracts (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 7.83pg/mL
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity Cross-reactivity observed with 1 or more available related molecules, < 50% cross-species reactivity observed with species tested
Product Type Human BMP-7 Quantikine ELISA Kit
Sample Type Bone Extracts, Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human BMP-7
For Use With (Application) ELISA
Gene ID (Entrez)
  • 655 (Human)
  • 12162 (Mouse)
  • 85272 (Rat)
Precision
  • Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested on one plate to assess intra-assay precision
  • Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in separate assays to assess inter-assay precision
Quantity 1 Pack
Sample Volume Cell Culture Supernates (50uL),Bone Extracts (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1 OP1, Osteogenic protein 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.