missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-6 Recombinant Protein, PeproTech®

Product Code. 17800963
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17800963

Brand: Gibco™ 120061MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

120-06-1MG will be provided as 2 x 500 μg (120-06-500UG). Recombinant Human BMP-6 is a 26.2 kDa homodimeric glycoprotein consisting of two 117 amino acid subunits, which correspond to amino acid residues 397 to 513 of the full-length BMP-6 precursor. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

BMP6 is a member of bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, Bmp6 has a proposed role in early development. In addition, the fact that BMP6 is closely related to BMP5 and BMP7 has lead to speculation of possible bone inductive activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22004
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 654
Molecular Weight (g/mol) 26.2
Name Human BMP-6
Quantity 2 x 500 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BMP; BMP 6; BMP6; BMP-6; Bone morphogenetic protein; Bone morphogenetic protein 6; D13Wsu115e; vegetal related growth factor (TGFB-related); vegetal-related (TGFB related) cytokine; VG-1-R; VG-1-related protein; Vg1-related sequence; Vgr; Vgr1; VGR-1
Gene Symbol BMP6
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Sequence VSSASDYNSS ELKTACRKHE LYVSFQDLGW QDWIIAPKGY AANYCDGECS FPLNAHMNAT NHAIVQTLVH LMNPEYVPKP CCAPTKLNAI SVLYFDDNSN VILKKYRNMV VRACGCH
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.