missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-6 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF507
This item is not returnable.
View return policy
Description
BMP-6 Polyclonal specifically detects BMP-6 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| BMP-6 | |
| Polyclonal | |
| Biotin | |
| P22004 | |
| BMP6 | |
| E. coli-derived recombinant human BMP-6 (R&D Systems, Catalog # 507-BP) Gln382-His513 Accession # P22004 | |
| 50 μg | |
| Primary | |
| Detects human BMP-6 in ELISAs and Western blots. In sandwich immunoassays, less than 4% cross-reactivity with recombinant human (rh) BMP-5 is observed and less than 0.5% cross-reactivity with rhBMP-2, rhBMP-4, rhBMP-7, rhBMP-8, rhTGF-beta 1, rhTGF-beta 2, and rhTGF-beta 3 is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, Vgr-1, VGR1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 654 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction