missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-4 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF757
This item is not returnable.
View return policy
Description
BMP-4 Polyclonal specifically detects BMP-4 in Human samples. It is validated for Western Blot.
Specifications
| BMP-4 | |
| Polyclonal | |
| Biotin | |
| P12644 | |
| BMP4 | |
| Mouse myeloma cell line NS0-derived recombinant human BMP-4 Ser293-Arg408 Accession # P12644 | |
| 50 μg | |
| Primary | |
| Detects human BMP-4 in Western blots. In Western blots, approximately 35% cross-reactivity with recombinant human (rh) BMP-2 is observed and 10% cross-reactivity with rhBMP-5 and rhBMP-6 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 652 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction