missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£169.00 - £459.00
Specifications
| Antigen | BMP-4 |
|---|---|
| Clone | 1050731 |
| Dilution | Immunocytochemistry 8 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723217
![]() |
R&D Systems
MAB7572-SP |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772517
![]() |
R&D Systems
MAB7572-100 |
100 μg |
£459.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-4 Monoclonal antibody specifically detects BMP-4 in Human samples. It is validated for ImmunocytochemistrySpecifications
| BMP-4 | |
| Immunocytochemistry 8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME | |
| E. coli-derived human BMP-4 protein, Lys303-Arg408, Accession # P12644 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1050731 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P12644 | |
| 652 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title