Learn More
Gibco™ Human BMP-4, Animal-Free Recombinant Protein, PeproTech®
Description
AF-120-05ET-1MG will be provided as 2 x 500 μg (AF-120-05ET-500UG). PeproTech's E.coli-derived BMP-4 is a fully active homodimeric protein consisting of two 107 amino acid subunits, which correspond to amino acids 303-408 of the full length BMP-4 precursor, and includes a N-terminal Met residue. The calculated molecular weight of Recombinant Human BMP-4 (E.coli-derived) is 24.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P12644 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 652 |
| Molecular Weight (g/mol) | 24.2 kDa |
| Name | Human BMP-4, Animal-Free |
| Purification Method | Purified |
| Quantity | 500 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.