missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-3 Recombinant Protein, PeproTech®

Product Code. 17830403
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17830403

Brand: Gibco™ 12024B10UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BMP-3 is a disulfide-linked homodimeric protein that corresponds to residues 361 to 472 of the 472 amino acid BMP-3 precursor protein. The calculated molecular weight of Recombinant Human BMP-3 is 25.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The BMPs belong to the TGF-Beta superfamilyBMPs stimulate the production of bone matrix proteins and thereby alter stromal cell and osteoclast proliferation, a key step in bone marrow development. In addition to promoting ectopic bone and cartilage development, BMPs regulate the growth, differentiation, chemotaxis, proliferation, and apoptosis of various cell types, including mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells. BMPs also play a role in dorsal/ventral patterning. BMP3, highly expressed in lung, ovary and small intestine, participates in cartilage and bone formation. BMP3 and BMP2 genes map to conserved regions between human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12645
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 651
Molecular Weight (g/mol) 25.2 kDa
Name Human BMP-3
Quantity 10 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 9130206H07; 9530029I04Rik; BMP; Bmp3; Bmp-3; BMP3A; BMP-3 A; Bone morphogenetic protein; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); bone morphogenetic protein 3 A; bone morphogenetic protein-3; osteogenin; PBMP3
Common Name BMP-3
Gene Symbol BMP3
Biological Activity Determined by its ability to inhibit BMP-2-induced alkaline phosphatase production by ATDC-5 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQWIEPRNCA RRYLKVDFAD IGWSEWIISP KSFDAYYCSG ACQFPMPKSL KPSNHATIQS IVRAVGVVPG IPEPCCVPEK MSSLSILFFD ENKNVVLKVY PNMTVESCAC R
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.