missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-2 Recombinant Protein

Recombinant Human Bone Morphogenetic Protein 2 (BMP2) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC7146

Code : 50

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10318624

  • £652.00 / Each
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Human BMP-2 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

BMP2 is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. BMP2 acts as a disulfide-linked homodimer and induces bone and cartilage formation.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent induction of alkaline phosphatase in mouse chondrogenic cell line ATDC5
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice.

Specifications

Specifications

NM001200
548717
Sequential Chromatography
BMP-2, BMP-2, MGC114605, xBMP-2, xbmp2
ED50 range = 0.1-1.0 mg⁄mL
Store lyophilized human BMP-2 at 2 to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤ –20°C. Avoid repeated freeze⁄thaw cycles.
< 0.1 ng/μg
10 μg
Wet Ice
E. coli
Recombinant, Ligand
BMPs (Bone Morphogenetic Proteins) & Related Factors
Gibco™
Cell Culture
26 kDa
25μg
Stem Cells
Human
Carrier-Free
mouse chondrogenic cell line ATDC5
Human
1.0 x 104 - 1.0 x103 units⁄mg
Growth Factors & Receptors
Lyophilized
Stem Cell Research, Bone Research
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.