missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-15/GDF-9B Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2925-SP
This item is not returnable.
View return policy
Description
BMP-15/GDF-9B Monoclonal specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BMP-15/GDF-9B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O95972 | |
| BMP15 | |
| E. coli-derived recombinant human BMP-15/GDF-9B Gln268-Arg392 Accession # O95972 | |
| 25 μg | |
| Primary | |
| Detects human BMP-15/GDF-9B in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human BMP-1.1, -2, -3, -3b, -4, -5, -6, -7, -8, or -10 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 317809 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP15, BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9210 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction