missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-10 Recombinant Protein, PeproTech®

Product Code. 17850133
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
This item is not returnable. View return policy

Product Code. 17850133

Brand: Gibco™ 120401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BMP-10 is a 24.4 kDa homodimeric disulfide-linked protein consisting of two 108 amino acid subunits, which correspond to amino acid residues 317 to 424 of the full-length BMP-10 precursor. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-beta superfamily of structurally related signaling proteins. Members of this superfamily are widely distributed throughout the body and are involved in diverse physiological processes during both pre- and postnatal life. BMP-10 plays a crucial role in the development of the embryonic heart by acting to stimulate and maintain cardiomyocyte proliferation. It can signal through various receptor complexes usually containing BMPR-1A, BMPR-1B, ALK1, ALK3, or ALK6. The interaction of BMP-10 with its specific receptors can induce signaling initiated by the phosphorylation of SMAD transcription factors, including SMAD1, SMAD5, or SMAD8, but can also induce SMAD independent processes. BMP-10 is structurally related to BMP-9, and both can inhibit endothelial cell proliferation and migration.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95393
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 27302
Molecular Weight (g/mol) 24.4 kDa
Name Human BMP-10
Quantity 1 mg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BMP; BMP10; BMP-10; bmp10 {ECO:0000312; bmp10b; Bone morphogenetic protein; bone morphogenetic protein 10; EMBL:BAE16990.1, ECO:0000312; RGD:1562986}; si:dkey-13n15.5
Common Name BMP-10
Gene Symbol BMP10
Biological Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 4.0-6.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence NAKGNYCKRT PLYIDFKEIG WDSWIIAPPG YEAYECRGVC NYPLAEHLTP TKHAIIQALV HLKNSQKASK ACCVPTKLEP ISILYLDKGV VTYKFKYEGM AVSECGCR
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.