missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-10 Propeptide Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £460.00
Specifications
| Antigen | BMP-10 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707237
![]() |
R&D Systems
AF3956-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18022792
|
R&D Systems
AF3956 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-10 Polyclonal specifically detects BMP-10 in Human samples. It is validated for Western Blot.Specifications
| BMP-10 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| BMP10, BMP-10, bone morphogenetic protein 10, MGC126783 | |
| BMP10 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O95393 | |
| 27302 | |
| Mouse myeloma cell line NS0-derived recombinant human BMP-10 propeptide (R&D Systems, Catalog # 3956-BP) Ser20-Arg313 Accession # O95393 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human BMP-10 propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with mature recombinant human BMP-10 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title