missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human BMP-1/PCP (NP_006120) VersaClone cDNA
Click to view available options
:
10 μg
Description
- Also known as: PCP; TLD; OI13; PCP2; PCOLC
- BMP-1 is a zinc protease of the astacin family.
- BMP-1 plays a key role in formation of extracellular matrix (ECM) by converting precursor proteins into their mature and functional forms.
- The precursor proteins identified as substrates for BMP-1 include collagens, biglycan, laminin 5, dentin matrix protein-1, and lysyl oxidase.
- Alternatively spliced transcripts encoding different proteins have been described.
- The BMP-1/PCP insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | BMP-1/PCP |
| Accession Number | NP_006120 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2974 bp |
| Gene ID (Entrez) | 649 |
| Gene Alias | BMP1, BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction