missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMAL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB105601-100
This item is not returnable.
View return policy
Description
BMAL1 Monoclonal specifically detects BMAL1 in Human samples. It is validated for ELISA.
Specifications
| BMAL1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| ARNTL, aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1c, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, JAP3, Member of PAS protein 3, member of PAS superfamily 3, MOP3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3, PASD3MGC47515, TIC | |
| E.coli-derived recombinant human BMAL1, Met105-Glu303, Accession # O00327 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 1029533 | |
| ELISA | |
| O00327 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 406 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction