missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BLOC1S1 (aa 33-147) Control Fragment Recombinant Protein

Product Code. 30205307
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205307

Brand: Invitrogen™ RP91631

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54395 (PA5-54395. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

BLOC1S1 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell'Angelica, 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78537
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2647
Name Human BLOC1S1 (aa 33-147) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI839753; biogenesis of lysosomal organelles complex 1 subunit 1; biogenesis of lysosomal organelles complex-1, subunit 1; biogenesis of lysosome-related organelles complex 1 subunit 1; biogenesis of lysosome-related organelles complex 1 subunit 1-like; biogenesis of lysosome-related organelles complex-1, subunit 1; BLOC subunit 1; BLOC-1 subunit 1; BLOC1S1; BLOS1; BORCS1; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 1; GCN5 general control of amino acid synthesis-like 1; GCN5 general control of amino-acid synthesis 5-like 1; Gcn5l1; GCN5-like protein 1; general control of amino acid synthesis, yeast homolog-like 1; general control of amino acid synthesis-like 1; MICoA; MTA1-interacting coactivator; Protein RT14; RT14; si:dkey-102c8.4; Unknown (protein for MGC:137062)
Common Name BLOC1S1
Gene Symbol Bloc1s1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLKEHQAKQNERKELQEKRRREAITAATCLTEALVDHLNVGVAQAYMNQRKLDHEVKTLQVQAAQFAKQTGQWIGMVENFNQALKEIGDVENWARSIELDMRTIATALEYVYKGQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.