missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human BIN1 - Ready-To-Use ELISA Kit (Colorimetric)

Product Code. 30056749
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30056749 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30056749 Supplier Novus Biologicals™ Supplier No. NBP331152

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.56 - 100 pg/mL (example only; lot dependent)
Assay Sensitivity 0.51 pg/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Tissue Homogenates, Biological Fluids
For Use With (Application) ELISA
Gene ID (Entrez) 274
Gene Symbol BIN1
Quantity 1 Kit
Research Discipline Cell Cycle and Replication, Tumor Suppressors
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym AMPH2, Amphiphysin II, Amphiphysin-like protein, AMPHLDKFZp547F068, box dependant MYC interacting protein 1, bridging integrator 1Box-dependent myc-interacting protein 1, MGC10367, myc box-dependent-interacting protein 1, SH3P9
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.