missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BHE23 (aa 163-225) Control Fragment Recombinant Protein

Product Code. 30198137
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198137

Brand: Invitrogen™ RP95507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56510 (PA5-56510. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the basic helix-loop-helix transcription factor family. Members of this family contain two highly conserved and functionally distinct domains: the basic domain targets sequence-specific DNA binding, while the helix-loop-helix domain facilitates protein interaction. Studies of a related gene in mouse suggest that the encoded protein may function as a transcriptional repressor in the pancreas and brain, and that it is required for normal retinal function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NDY6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 128408
Name Human BHE23 (aa 163-225) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A930001L02Rik; bA305P22.3; basic helix-loop-helix domain containing, class B, 4; basic helix-loop-helix domain containing, class B4; basic helix-loop-helix family member e23; basic helix-loop-helix family, member e23; BETA4; beta-cell E-box transactivating factor 4; BHLHB4; BHLHE23; Class B basic helix-loop-helix protein 4; class E basic helix-loop-helix protein 23; H20; RGD1559760
Common Name BHE23
Gene Symbol Bhlhe23
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RLVAFLNQGQGLAAPVNAAPLTPFGQATVCPFSAGAALGPCPDKCAAFSGTPSALCKHCHEKP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.