missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Betacellulin Recombinant Protein, PeproTech®

Product Code. 17891423
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17891423

Brand: Gibco™ 10050100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Betacellulin is a 9.0 kDa monomeric protein, containing 80 amino acid residues, which comprises the mature EGF-homologous portion of theBetacellulin protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35070
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 685
Molecular Weight (g/mol) 9 kDa
Name Human Betacellulin
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin
Common Name Betacellulin
Gene Symbol BTC
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine Balb/3T3 cells is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 10^7 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence DGNSTRSPET NGLLCGDPEE NCAATTTQSK RKGHFSRCPK QYKHYCIKGR CRFVVAEQTP SCVCDEGYIG ARCERVDLFY
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.