missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human beta-NGF DuoSet ELISA
Click to view available options
Quantity:
15 x 96-well Plates
Unit Size:
1 set
Description
The Human beta-NGF DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human beta-NGF.
Specifications
Specifications
| Quantity | 15 x 96-well Plates |
| Conjugate | Biotin |
| Assay Range | 31.20 to 2000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 4803 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction