missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-NGF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB256-SP
This item is not returnable.
View return policy
Description
beta-NGF Monoclonal specifically detects beta-NGF in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| beta-NGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Beta nerve growth factor precursor, beta-nerve growth factor, betaNGF, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFB, NGFBMGC161428 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4803 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization, ELISA | |
| 25623 | |
| Neutralization 0.1-0.4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CAA36832 | |
| NGF | |
| Mouse myeloma cell line NS0-derived recombinant human beta-NGF Ser122-Ala241 Accession # CAA36832 | |
| 25 μg | |
| Primary | |
| Detects human beta-NGF in ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) beta-NGF and recombinant rat (rr) beta-NGF is observed and less than 5% cross-reactivity with recombinant human (rh) BDNF, rhNT-3, and rhNT-4 is observed. This antibody neutralizes the bioactivity of rh beta-NGF, rm beta-NGF, and rr beta-NGF. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction