missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta IG-H3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2935
Additional Details : Weight : 0.00970kg
Description
Beta Ig-h3/TGFBI Polyclonal specifically detects Beta Ig-h3/TGFBI in Human samples. It is validated for Western Blot.Specifications
Beta Ig-h3/TGFBI | |
Polyclonal | |
Unconjugated | |
Q15582 | |
TGFBI | |
Mouse myeloma cell line NS0-derived recombinant human beta IG-H3 Gly24-His683 Accession # Q15582 | |
100 μg | |
Primary | |
Detects human beta IG-H3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse beta IG-H3 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
beta IGH3, Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, TGFBI, TGFBIP, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3 | |
Goat | |
Affinity Purified | |
RUO | |
7045 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only