missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta IG-H3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £523.00
Specifications
| Antigen | Beta Ig-h3/TGFBI |
|---|---|
| Clone | 348506 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753399
![]() |
R&D Systems
MAB2935-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18192541
![]() |
R&D Systems
MAB2935 |
500 μg |
£523.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Beta Ig-h3/TGFBI Monoclonal specifically detects Beta Ig-h3/TGFBI in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| Beta Ig-h3/TGFBI | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta IGH3, Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, TGFBI, TGFBIP, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3 | |
| TGFBI | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 348506 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q15582 | |
| 7045 | |
| Mouse myeloma cell line NS0-derived recombinant human beta IG-H3 Gly24-His683 Accession # Q15582 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human beta IG-H3 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse beta IG-H3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title