missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-Glucuronidase/GUSB Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6144-SP
This item is not returnable.
View return policy
Description
beta-Glucuronidase/GUSB Polyclonal specifically detects beta-Glucuronidase/GUSB in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| beta-Glucuronidase/GUSB | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-D-glucuronidase, beta-G1, beta-glucuronidase, BG, EC 3.2.1.31, FLJ39445, glucuronidase, beta, MPS7 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2990 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| AAA52561 | |
| GUSB | |
| Mouse myeloma cell line NS0-derived recombinant human beta-Glucuronidase/GUSB Met1-Thr651 Accession # AAA52561 | |
| 25 μg | |
| Primary | |
| Detects human beta-Glucuronidase/GUSB in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction