missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Beta Endorphin ELISA Kit

Product Code. 18077454
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18077454

Brand: Invitrogen™ EEL031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Beta endorphin is an opioid neuropeptide that is produced by neurons of the pituitary gland and released into the circulatory system, or produced by the hypothalamus, and released into the brain and spinal cord through nerve fibers. The precursor polypeptide for beta endorphin is pro-opiomelanocortin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01189
Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5443
Gene Symbol POMC
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Beta Endorphin
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.