missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-Defensin 4/2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF2758
This item is not returnable.
View return policy
Description
beta-Defensin 4/2 Polyclonal specifically detects beta-Defensin 4/2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| beta-Defensin 4/2 | |
| Polyclonal | |
| Unconjugated | |
| O15263 | |
| DEFB4A | |
| E. coli-derived recombinant human beta-Defensin 4/2 Gly24-Pro64 Accession # O15263 | |
| 100 μg | |
| Primary | |
| Detects human beta-Defensin 4/2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| BD-2, betaDefensin 2, Beta-defensin 2, DEFB2, DEFB-2, DEFB2DEFB102, DEFB4A, DEFB4B, DEFB4beta-defensin 4A, Defensin, beta 2Skin-antimicrobial peptide 1, defensin, beta 4, defensin, beta 4A, HBD-2, SAP-1, SAP1BD-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1673 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction